iFluor® 440, Succinimidyl Ester fluorescent probe

Toimittaja: AAT BIOQUEST
Varoitus

71041 71519 71569 1041
AATB71041EA 114 EUR
AATB71041 AATB71519 AATB71569 AATB1041
iFluor® 440, Succinimidyl Ester fluorescent probe
iFluor®

AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.


  • pH-insensitive
  • Azide reactive
  • High photostability
  • Brighter signal

ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR