Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (mFluor™ Violet 500)

Toimittaja: AAT BIOQUEST
10033100 10033101
AATB10033100EA 371 EUR
AATB10033100 AATB10033101
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (mFluor™ Violet 500)
Vasta-aineet
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Varoitus: For research use only.

Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: mFluor™ Violet 500
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR