Acriflavine hydrochloride fluorescent dye, Sigma-Aldrich®

Toimittaja: SIGMA ALDRICH MICROSCOPY
Vaara

Synonyms: 3,6-Diamino-10-methylacridinium chloride hydrochloride, Euflavine

M-Clarity™ quality level = MQ200

A8251-100G A8251-10G A8251-25G
A8251-100GEA 269 EUR
A8251-100G A8251-10G A8251-25G
Acriflavine hydrochloride fluorescent dye, Sigma-Aldrich®
Acriflavine hydrochloride

In living mice, acriflavine hydrochloride has been used in the confocal laser endomicroscopy to check the crypt structure in mucosa. It has also been used in Neurospora strain selection by checking the acriflavine resistance.


Acriflavine hydrochloride is a fluorescent dye. It is used as a topical contrast agent. It mainly stains nuclei and is applied to tissues before microscopy.


A mixture of 3,6-diamino-10-methylacridinium (acriflavin) and 3,6-diaminoacridine (proflavin).

Kaava: C₂₇H₂₈Cl₄N₆
Sulamispiste: 260 °C (haj. lämpötila, lit.)
Storage Temperature: Room Temperature
MDL numero: MFCD00069039
CAS-numero: 8063-24-9
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86ceac016d054f0612eba595de224ddf42fb93f0&vid=e2b35ebcb24d1ab716de8f633ac4f9612a4782e2&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance (Colour) Faint Brown to Brown and Orange to Dark Orange and Red to Very Dark Red
Appearance (Form) Powder
Solubility (Colour) Orange to Brown
Solubility (Turbidity)
1 mg/ml, H₂O
Clear
Water (by Karl Fischer) ≤13 %
Infrared spectrum Conforms to Structure
% Acriflavine HCL content 10 - 50
% Proflavine HCL 25 - 70
Recommended Retest Period 3 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR