Decontamination Solution, DNA/RNA-ExitusPlus™ IF (Indicator-free)
Toimittaja: PanReac AppliChem
A7409.0100 A7409.0500 A7409.1000RF A7409.5000
A7409.0100EA 66.9 EUR
A7409.0100 A7409.0500 A7409.1000RF A7409.5000
Decontamination Solution, DNA/RNA-ExitusPlus™ IF (Indicator-free)
Puhdistusaineet Dekontaminaatioliuokset nukleiinihapoille
Decontamination solution for the removal of DNA and RNA from surfaces.
  • All components are readily not harmful or toxic for humans
  • Doesn't contain aggressive mineral acids or alkaline substances
  • No toxic fumes
  • Contains low concentration of alcohol