Natriumkloridi ≥98.5% for molecular biology

Toimittaja: PanReac AppliChem

Synonyms: Saline, NaCl, Sea salt, Halite

A2942.1000 A2942.5000
A2942.1000EA 54.2 EUR
A2942.1000 A2942.5000
Natriumkloridi ≥98.5% for molecular biology
Natriumkloridi
Kaava: NaCl
MW: 58.44 g/mol
Kiehumispiste: 1413
Sulamispiste: 801
Tiheys: 2.16 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003477
CAS-numero: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Testitulosten erittely

DNases/RNases/Proteases not detectable
Assay (titr.) min. 99.5 %
pH (5 %; H2O; 20°C) 5.0 - 8.0
Heavy metals (as Pb) max. 0.0005 %
Bromide max. 0.005 %
Iodide max. 0.001 %
Phosphate max. 0.001 %
Sulfate max. 0.01 %
Ba max. 0.001 %
Ca max. 0.002 %
Fe max. 0.0001 %
K max. 0.01 %
MG max. 0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR