ACCUSPIN™ Centrifuge Tubes, Sterile, Polypropylene
Toimittaja: SIGMA ALDRICH MICROSCOPY
A1805-20EA A2055-10EA
A1805-20EAEA 171 EUR
A1805-20EA A2055-10EA
ACCUSPIN™ Centrifuge Tubes, Sterile, Polypropylene
Putket Sentrifugiputket
ACCUSPIN™ Tubes are radiation sterilised tubes fitted with a high density polyethylene barrier.
  • Sterile, irradiated
  • Ideal for hematology and histology applications
ACCUSPIN™ tubes is intended for use with the density gradient separation medium Histopaque™ -1077:
- in the isolation of lymphocytes and other mononuclear cells
- isolation of cord blood–derived endothelial progenitor cells
- isolate mononuclear cell layer from bone marrow samples
- suitable for the purification of human bone marrow aspirates for the investigation of the therapeutic value ofMore Product Information