Sodium-tert-butanolate ≥97%

Toimittaja: Thermo Fisher Scientific
Vaara

A19942.22 A19942.0E A19942.36
A19942.22EA 127 EUR
A19942.22 A19942.0E A19942.36
Sodium-tert-butanolate ≥97%
Sodium-tert-butanolate

Purity: 97% CAS No.: 865-48-5 Molecular Formula: C4H9NaO Formula Weight: 96.11 Odour: Odourless Beilstein Reference: 3654215 UN No.: 3206 MDL No.: MFCD00040575

Kaava: C₄H₉NaO
Tiheys: 0,785
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00040575
CAS-numero: 865-48-5
EINECS: 212-741-9
UN: 3206
ADR: 4.2,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR