N,N-Diethylethylenediamine 99%

Toimittaja: Thermo Fisher Scientific
Vaara

A19636.22 A19636.36
A19636.22EA 85.2 EUR
A19636.22 A19636.36
N,N-Diethylethylenediamine 99%
N,N-Diethylethylenediamine

Purity: 99% CAS No.: 100-36-7 Molecular Formula: (CH3CH2)2NCH2CH2NH2 Formula Weight: 116.21 Flash Point: 30°(86°F) Refractive Index: 1.4360 Melting Point: -70° Boiling Point: 145-147° Beilstein Reference: 605299 UN No.: 2685 MDL No.: MFCD00008176

Kaava: C₆H₁₆N₂
Kiehumispiste: 145…147 °C
Sulamispiste: < –70 °C
Tiheys: 0,825
Leimahduspiste: 30 °C (86 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008176
CAS-numero: 100-36-7
EINECS: 202-844-7
UN: 2685
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR