3-Bromo-2-(bromomethyl)propionic acid 98%

Toimittaja: Thermo Fisher Scientific
Vaara

A19630.14 A19630.22 A19630.06
A19630.14EA 639 EUR
A19630.14 A19630.22 A19630.06
3-Bromo-2-(bromomethyl)propionic acid 98%
3-Bromo-2-(bromomethyl)propionic acid

Purity: 98% CAS No.: 41459-42-1 Molecular Formula: (BrCH2)2CHCO2H Formula Weight: 245.91 Melting Point: 98-101° UN No.: UN3261 MDL No.: MFCD00010643

Kaava: C₄H₆Br₂O₂
Sulamispiste: 98…101 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010643
CAS-numero: 41459-42-1
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR