(4-Bromifenyyli)asetoni ≥98%

Toimittaja: Thermo Fisher Scientific

A19028.14 A19028.06
A19028.14EA 541 EUR
A19028.14 A19028.06
(4-Bromifenyyli)asetoni ≥98%
(4-Bromifenyyli)asetoni

Purity: 98% CAS No.: 6186-22-7 Molecular Formula: C9H9BrO Formula Weight: 213.07 MDL No.: MFCD00210401

Kaava: C₉H₉BrO
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00210401
CAS-numero: 6186-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR