Kaliumhydroksidi ≥85%, pellets

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: Kalilipeä

A18854.36 A18854.0E
A18854.36EA 69.1 EUR
A18854.36 A18854.0E
Kaliumhydroksidi ≥85%, pellets
Kaliumhydroksidi

Purity: 85% CAS No.: 1310-58-3 Molecular Formula: KOH Formula Weight: 56.11 Odour: Odourless pH: 13,5 Melting Point: 360° Boiling Point: 1320-1324° Merck Reference: 14,7640 UN No.: 1813 MDL No.: MFCD00003553

Kaava: KOH
MW: 56.11 g/mol
Kiehumispiste: 1320…1324 °C
Sulamispiste: 360
Tiheys: 2.044
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003553
CAS-numero: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8 II
REACH: 01-2119487136-33
Merck Index: 14,7640

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR