Zinc chromate

Toimittaja: Thermo Fisher Scientific
Vaara

A18178.30 A18178.0B
A18178.30EA 164 EUR
A18178.30 A18178.0B
Zinc chromate
Zinc chromate

CAS No.: 13530-65-9 Molecular Formula: ZnCrO4 Formula Weight: 181.37 UN No.: UN3077 MDL No.: MFCD00036281

Kaava: ZnCrO₄
Tiheys: 3,4
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00036281
CAS-numero: 13530-65-9
EINECS: 236-878-9
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR