Oleyl alcohol tech. 80-85%, Technical Grade

Toimittaja: Thermo Fisher Scientific
Varoitus

A18018.22 A18018.36
A18018.22EA 65 EUR
A18018.22 A18018.36
Oleyl alcohol tech. 80-85%, Technical Grade
Oleyl alcohol

Purity: 80-85% CAS No.: 143-28-2 Molecular Formula: C18H36O Formula Weight: 268.48 Flash Point: 180°(356°F) Physical Form: Liquid Density: 0.849 Refractive Index: 1.4600 Appearance: Colourless Melting Point: ca 6° Boiling Point: 206-207°/13mm Beilstein Reference: 1723962 Merck Reference: 14,6831 MDL No.: MFCD00002993

Kaava: C₁₈H₃₆O
Kiehumispiste: 206…207 °C (13 mmHg)
Sulamispiste: ∼ 6 °C
Tiheys: 0,849
Leimahduspiste: 180 °C (356 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002993
CAS-numero: 143-28-2
EINECS: 205-597-3
Merck Index: 14,6831

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR