Tris(3-methoxyphenyl)phosphine ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

A17949.03 A17949.06
A17949.03EA 140 EUR
A17949.03 A17949.06
Tris(3-methoxyphenyl)phosphine ≥98%
Tris(3-methoxyphenyl)phosphine

Purity: 98% CAS No.: 29949-84-6 Molecular Formula: C21H21O3P Formula Weight: 352.37 Melting Point: 113-115° MDL No.: MFCD00008386

Kaava: C₂₁H₂₁O₃P
Sulamispiste: 113…115 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008386
CAS-numero: 29949-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR