L(-)-Mannoosi ≥99%

Toimittaja: Thermo Fisher Scientific

A17722.01 A17722.03 A17722.06
A17722.01EA 113 EUR
A17722.01 A17722.03 A17722.06
L(-)-Mannoosi ≥99%
L(-)-Mannoosi

Purity: 99% CAS No.: 10030-80-5 Molecular Formula: C6H12O6 Formula Weight: 180.16 Melting Point: 129-131° Beilstein Reference: 1724628

Kaava: C₆H₁₂O₆
Kiehumispiste: 376 °C (1013 hPa)
Sulamispiste: 129…131 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00136021
CAS-numero: 10030-80-5
EINECS: 233-080-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR