Terbium triacetate hydrate 99%

Toimittaja: Thermo Fisher Scientific

A17388.06 A17388.14
A17388.06EA 328 EUR
A17388.06 A17388.14
Terbium triacetate hydrate 99%
Terbium triacetate hydrate

Purity: 99.99% CAS No.: 100587-92-6 Molecular Formula: Tb(OOCCH3)3.xH2O Formula Weight: 336.08(anhy) MDL No.: MFCD00149233

Kaava: C₆H₉O₆Tb·H₂O
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149233
CAS-numero: 100587-92-6
EINECS: 247-065-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR