Cyclohexylamine hydrochloride ≥98%

Toimittaja: Thermo Fisher Scientific
Vaara

A17308.22 A17308.36
A17308.22EA 64.3 EUR
A17308.22 A17308.36
Cyclohexylamine hydrochloride ≥98%
Cyclohexylamine hydrochloride

Purity: 98+% CAS No.: 4998-76-9 Molecular Formula: C6H13N.HCl Formula Weight: 135.64 Odour: Odourless Melting Point: 206-209° Beilstein Reference: 3613002 MDL No.: MFCD00050540

Kaava: C₆H₁₃N
Sulamispiste: 206…209 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00050540
CAS-numero: 4998-76-9
EINECS: 225-661-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR