D-(-)-Lyxose ≥99%

Toimittaja: Thermo Fisher Scientific

A17007.0025 AV17007.0025 A17007.06 A17007.14
ABCRA17007.0025EA 0 EUR
ABCRA17007.0025 ABCRAV17007.0025 A17007.06 A17007.14
D-(-)-Lyxose ≥99%
D-(-)-Lyxose

Purity: 99% CAS No.: 1114-34-7 Molecular Formula: C5H10O5 Formula Weight: 150.13 Melting Point: 109-115° Beilstein Reference: 1723083 Merck Reference: 14,5641 MDL No.: MFCD00064362

Kaava: C₅H₁₀O₅
Sulamispiste: 109…115 °C
Tiheys: 1,545
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00064362
CAS-numero: 1114-34-7
EINECS: 214-212-8
Merck Index: 14,5641

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR