Benzyltrimethylammonium chloride ≥97%

Toimittaja: Thermo Fisher Scientific
Vaara

A16759.22 A16759.36
A16759.22EA 61.5 EUR
A16759.22 A16759.36
Benzyltrimethylammonium chloride ≥97%
Benzyltrimethylammonium chloride

Purity: 97% CAS No.: 56-93-9 Molecular Formula: C6H5CH2N(CH3)3Cl Formula Weight: 185.70 Refractive Index: 1.4440 Odour: Odourless pH: 6-8 Melting Point: ca 238° dec. Beilstein Reference: 3917255 MDL No.: MFCD00011782

Kaava: C₁₀H₁₆ClN
Tiheys: 1,07 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011782
CAS-numero: 56-93-9
EINECS: 200-300-3
UN: 2811
ADR: 6.1,III

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR