Natriumkloridi 0,9% in aqueous solution, sterile

Toimittaja: PanReac AppliChem

Synonyms: Saline, NaCl, Sea salt, Halite

A1671.0100 A1671.0250 A1671.0500 A1671.1000
A1671.0100EA 69.9 EUR
A1671.0100 A1671.0250 A1671.0500 A1671.1000
Natriumkloridi 0,9% in aqueous solution, sterile
Natriumkloridi
Kaava: NaCl
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003477
CAS-numero: 7647-14-5
EINECS: 231-598-3

Order Now

Testitulosten erittely

pH (20 °C) 5,0 - 7,5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR