Kaliumbromidi ≥99%

Toimittaja: Thermo Fisher Scientific
Varoitus

A16339.0B A16339.0I A16339.22 A16339.30 A16339.36 A16339.A4
A16339.0BEA 176 EUR
A16339.0B A16339.0I A16339.22 A16339.30 A16339.36 A16339.A4
Kaliumbromidi ≥99%
Kaliumbromidi

Purity: 99+% CAS No.: 7758-02-3 Molecular Formula: KBr Formula Weight: 119.01 Refractive Index: 1.559 Odour: Odourless Boiling Point: 1435° Merck Reference: 14,7618 MDL No.: MFCD00011358

Kaava: KBr
Kiehumispiste: 1435 °C
Sulamispiste: 730 °C
Tiheys: 2,75
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011358
CAS-numero: 7758-02-3
EINECS: 231-830-3
Merck Index: 14,7618

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR