Rauta(III)kloridihexahydraatii ≥97%

Toimittaja: Thermo Fisher Scientific
Vaara

A16231.0C A16231.0E A16231.36
A16231.0CEA 1800 EUR
A16231.0C A16231.0E A16231.36
Rauta(III)kloridihexahydraatii ≥97%
Rauta(III)kloridihexahydraatii

Purity: 97% CAS No.: 10025-77-1 Molecular Formula: Cl3Fe.6H2O Formula Weight: 270.30 (162.21anhy) Odour: Odourless pH: 1.8 Melting Point: 37° Boiling Point: 280-285° Merck Reference: 14,4019 UN No.: 3260 MDL No.: MFCD00149712

Kaava: FeCl₃·6H₂O
Kiehumispiste: 280…285 °C
Sulamispiste: 37 °C
Tiheys: 1,820
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149712
CAS-numero: 10025-77-1
EINECS: 231-729-4
UN: 3260
ADR: 8,III
Merck Index: 14,4019
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=808c75376e55f29296d58eb1e5a1e184039ce733&vid=6d820873011bee3ff401441544f7f85599e40626&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR