DL-Histidine monohydrochloride monohydrate 99%

Toimittaja: Thermo Fisher Scientific

A16165.06 A16165.14 A16165.22
A16165.06EA 73.3 EUR
A16165.06 A16165.14 A16165.22
DL-Histidine monohydrochloride monohydrate 99%
DL-Histidine monohydrochloride monohydrate

Purity: 99% CAS No.: 123333-71-1 Molecular Formula: C6H9N3O2.HCl.H2O Formula Weight: 209.63 (191.62anhy) Melting Point: ca 250° dec. MDL No.: MFCD00151028

Kaava: C₆H₉N₃O₂·HCl·H₂O
Sulamispiste: ∼ 250 °C (haj. lämpötila)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00151028
CAS-numero: 123333-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR