Chlorophyllin, coppered trisodium salt

Toimittaja: Thermo Fisher Scientific

A16155.06 A16155.14 A16155.22
A16155.06EA 81.4 EUR
A16155.06 A16155.14 A16155.22
Chlorophyllin, coppered trisodium salt
Chlorophyllin, coppered trisodium salt

CAS No.: 11006-34-1 Molecular Formula: C34H31CuN4Na3O6 Formula Weight: 724.15 Storage Temperature: 2°C to 8°C Odour: Odourless MDL No.: MFCD00012149

Kaava: C₃₄H₃₁CuN₄Na₃O₆
MDL numero: MFCD00012149
CAS-numero: 11006-34-1
EINECS: 234-242-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR