Acetamidine hydrochloride ≥97%

Toimittaja: Thermo Fisher Scientific
Varoitus

A16091.36 A16091.14 A16091.22
A16091.36EA 359 EUR
A16091.36 A16091.14 A16091.22
Acetamidine hydrochloride ≥97%

Purity: 97% CAS No.: 124-42-5 Molecular Formula: C2H6N2.HCl Formula Weight: 94.54 Melting Point: 168-175° Beilstein Reference: 3591762 Merck Reference: 14,44 MDL No.: MFCD00013016

Kaava: C₂H₇ClN₂
Sulamispiste: 168…175 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00013016
CAS-numero: 124-42-5
EINECS: 204-700-9
Merck Index: 14,44

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR