4-Fluori-2-metyylifenoli ≥98%

Toimittaja: Thermo Fisher Scientific
Vaara

A16032.06 A16032.14 A16032.22
A16032.06EA 112 EUR
A16032.06 A16032.14 A16032.22
4-Fluori-2-metyylifenoli ≥98%
4-Fluori-2-metyylifenoli

Purity: 98% CAS No.: 452-72-2 Molecular Formula: C7H7FO Formula Weight: 126.13 Flash Point: 85°(185°F) Refractive Index: 1.5165 Odour: Phenol-like Melting Point: 32-36° Boiling Point: 87°/14mm Beilstein Reference: 2041490 UN No.: 3261 MDL No.: MFCD00075088

Kaava: C₇H₇FO
Kiehumispiste: 87 °C (14 mmHg)
Sulamispiste: 32…36 °C
Leimahduspiste: 85 °C (185 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00075088
CAS-numero: 452-72-2
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR