tert-Butyl chloroacetate ≥98%

Toimittaja: Thermo Fisher Scientific
Vaara

A15833.14 A15833.22 A15833.36
A15833.14EA 69.8 EUR
A15833.14 A15833.22 A15833.36
tert-Butyl chloroacetate ≥98%
tert-Butyl chloroacetate

Purity: 98% CAS No.: 107-59-5 Molecular Formula: C6H11ClO2 Formula Weight: 150.61 Flash Point: 46°(115°F) Refractive Index: 1.4230 Odour: Ester-like Boiling Point: 155° Beilstein Reference: 1753006 Merck Reference: 14,1563 UN No.: 2929 MDL No.: MFCD00000930

Kaava: C₆H₁₁ClO₂
Kiehumispiste: 155 °C
Tiheys: 1,053
Leimahduspiste: 46 °C (115 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00000930
CAS-numero: 107-59-5
EINECS: 203-506-1
UN: 2929
ADR: 6.1,II
Merck Index: 14,1563

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR