2-Ethyl-4-methylimidazole 96%

Toimittaja: Thermo Fisher Scientific
Vaara

A15798.14 A15798.22
A15798.14EA 99 EUR
A15798.14 A15798.22
2-Ethyl-4-methylimidazole 96%
2-Ethyl-4-methylimidazole

Purity: 96% CAS No.: 931-36-2 Molecular Formula: C6H10N2 Formula Weight: 110.16 Flash Point: 137°(278°F) Refractive Index: 1.5 00 Melting Point: 45-49° Boiling Point: 292-295° Beilstein Reference: 1711 MDL No.: MFCD00005193

Kaava: C₆H₁₀N₂
Kiehumispiste: 292…295 °C
Sulamispiste: 45…49 °C
Tiheys: 0,973
Leimahduspiste: 137 °C (278 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00005193
CAS-numero: 931-36-2
EINECS: 213-234-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR