Trinatriumfosfaatti-12 H2O ≥97%

Toimittaja: Thermo Fisher Scientific
Vaara

A15713.0C A15713.0E A15713.36
A15713.0CEA 577 EUR
A15713.0C A15713.0E A15713.36
Trinatriumfosfaatti-12 H2O ≥97%
Trinatriumfosfaatti-12 H2O

Purity: 97% CAS No.: 10101-89-0 Molecular Formula: Na3O4P.12H2O Formula Weight: 380.12 (163.94anhy) Odour: Odourless pH: 12 Melting Point: ca 75° dec. Merck Reference: 14,8662 UN No.: 3262 MDL No.: MFCD00149198

Kaava: Na₃PO₄·12H₂O
Sulamispiste: ∼75 °C (haj. lämpötila)
Tiheys: 1,62
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149198
CAS-numero: 10101-89-0
EINECS: 231-509-8
ADR: 8,III
REACH: 01-2119489800-32-xxxx
Merck Index: 14,8662

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR