3-Thenoic acid ≥99%

Toimittaja: Thermo Fisher Scientific
Varoitus

A15712.14 A15712.06 A15712.22
A15712.14EA 634 EUR
A15712.14 A15712.06 A15712.22
3-Thenoic acid ≥99%
3-Thenoic acid

Purity: 99% CAS No.: 88-13-1 Molecular Formula: C5H4O2S Formula Weight: 128.15 Melting Point: 137-141° Beilstein Reference: 1994 Merck Reference: 14,9279 MDL No.: MFCD00005467

Kaava: C₅H₄O₂S
Sulamispiste: 137…141 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00005467
CAS-numero: 88-13-1
EINECS: 201-802-5
Merck Index: 14,9279

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR