3-Metyyli-2-sykloheksen-1-oni ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

A15704.09 A15704.14 A15704.18 A15704.22
A15704.09EA 174 EUR
A15704.09 A15704.14 A15704.18 A15704.22
3-Metyyli-2-sykloheksen-1-oni ≥98%
3-Metyyli-2-sykloheksen-1-oni

Purity: 98% CAS No.: 1193-18-6 Molecular Formula: C7H10O Formula Weight: 110.15 Flash Point: 68°(154°F) Refractive Index: 1.4940 Odour: Pleasant Boiling Point: 199-200° Beilstein Reference: 1560601 MDL No.: MFCD00001581

Kaava: C₇H₁₀O
Kiehumispiste: 199…200 °C
Sulamispiste: 198…199 °C
Tiheys: 0,971
Leimahduspiste: 68 °C (154 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00001581
CAS-numero: 1193-18-6
EINECS: 214-769-7
UN: 1224
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR