Natriumsulfaatti-10 H2O ≥99%

Toimittaja: Thermo Fisher Scientific

A15702.0B A15702.0G A15702.0I
A15702.0BEA 125 EUR
A15702.0B A15702.0G A15702.0I
Natriumsulfaatti-10 H2O ≥99%
Natriumsulfaatti-10 H2O

Purity: 99% CAS No.: 7727-73-3 Molecular Formula: Na2SO4.10H2O Formula Weight: 322.19 (142.04anhy) Odour: Odourless pH: 5.2-8.0 Melting Point: ca 32° Merck Reference: 14,8680 MDL No.: MFCD00149189

Kaava: Na₂SO₄·10H₂O
Kiehumispiste: 1429 °C (1013 hPa)
Sulamispiste: ∼32 °C
Tiheys: 1,460
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149189
CAS-numero: 7727-73-3
EINECS: 231-820-9
Merck Index: 14,8680

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR