α,α,α-Trifluori-2-joditolueeni ≥99%

Toimittaja: Thermo Fisher Scientific
Varoitus

A15698.14 A15698.22
A15698.14EA 99.4 EUR
A15698.14 A15698.22
α,α,α-Trifluori-2-joditolueeni ≥99%
α,α,α-Trifluori-2-joditolueeni

Purity: 99% CAS No.: 444-29-1 Molecular Formula: C7H4F3I Formula Weight: 272.01 Flash Point: 81°(177°F) Physical Form: Liquid Density: 1.936 Refractive Index: 1.5305 Sensitivity: Light Sensitive Appearance: Colourless to pale yellow Boiling Point: 198-199° Beilstein Reference: 2090038 MDL No.: MFCD00001041

Kaava: C₇H₄F₃I
Kiehumispiste: 198…199 °C
Tiheys: 1,936
Leimahduspiste: 81 °C (177 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00001041
CAS-numero: 444-29-1
EINECS: 207-147-1

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR