Ethyl malonyl chloride 95%

Toimittaja: Thermo Fisher Scientific
Vaara

A15616.06 A15616.14 A15616.22
A15616.06EA 133 EUR
A15616.06 A15616.14 A15616.22
Ethyl malonyl chloride 95%
Ethyl malonyl chloride

Purity: 95% CAS No.: 36239-09-5 Molecular Formula: C5H7ClO3 Formula Weight: 150.56 Storage Temperature: 2°C to 8°C Flash Point: 67°(152°F) Refractive Index: 1.4290 Boiling Point: 83°/22mm Beilstein Reference: 636215 UN No.: 3265 MDL No.: MFCD00000736

Kaava: C₅H₇ClO₃
Kiehumispiste: 83 °C (22 mmHg)
Tiheys: 1,189
Leimahduspiste: 67 °C (152 °F)
MDL numero: MFCD00000736
CAS-numero: 36239-09-5
EINECS: 252-934-5
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR