Methyl-4-bromocrotonate tech. 85%, Technical Grade

Toimittaja: Thermo Fisher Scientific
Vaara

A15582.18 A15582.09 A15582.30
A15582.18EA 387 EUR
A15582.18 A15582.09 A15582.30
Methyl-4-bromocrotonate tech. 85%, Technical Grade
Methyl-4-bromocrotonate

Purity: 85% CAS No.: 1117-71-1 Molecular Formula: C5H7BrO2 Formula Weight: 179.01 Storage Temperature: 2°C to 8°C Flash Point: 91°(195°F) Refractive Index: 1.5010 Boiling Point: 83-85°/13mm Beilstein Reference: 1745755 UN No.: 1760 MDL No.: MFCD00000246

Kaava: C₅H₇BrO₂
Kiehumispiste: 83…85 °C (13 mmHg)
Tiheys: 1,524
Leimahduspiste: 91 °C (195 °F)
Storage Temperature: Jääkaappi
MDL numero: MFCD00000246
CAS-numero: 1117-71-1
EINECS: 214-251-0
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR