Methyl cyanoacetate ≥99%

Toimittaja: Thermo Fisher Scientific
Varoitus

A15555.0B A15555.22 A15555.30 A15555.36
A15555.0BEA 276 EUR
A15555.0B A15555.22 A15555.30 A15555.36
Methyl cyanoacetate ≥99%
Methyl cyanoacetate

Purity: 99% CAS No.: 105-34-0 Molecular Formula: NCCH2CO2CH3 Formula Weight: 99.09 Flash Point: 110°(230°F) Refractive Index: 1.4185 Odour: Mild Melting Point: -23° Boiling Point: 204° Beilstein Reference: 773945 MDL No.: MFCD00001939

Kaava: C₄H₅NO₂
Kiehumispiste: 204 °C
Sulamispiste: –23 °C
Tiheys: 1,127
Leimahduspiste: 110 °C (230 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00001939
CAS-numero: 105-34-0
EINECS: 203-288-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR