Hexaamminecobalt trichloride 99%

Toimittaja: Thermo Fisher Scientific
Varoitus

A15470.14 A15470.22 A15470.36
A15470.14EA 114 EUR
A15470.14 A15470.22 A15470.36
Hexaamminecobalt trichloride 99%
Hexaamminecobalt trichloride

Purity: 99% CAS No.: 10534-89-1 Molecular Formula: H18Cl3CoN6 Formula Weight: 267.48 Odour: Odourless Merck Reference: 14,4675 MDL No.: MFCD00036304

Kaava: H₁₈CoCl₃N₆
Tiheys: 1,71
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00036304
CAS-numero: 10534-89-1
EINECS: 234-103-9
Merck Index: 14,4675

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR