4-(Trifluoromethoxy)benzenesulphonyl chloride ≥98%

Toimittaja: Thermo Fisher Scientific
Vaara

A15467.03 A15467.06 A15467.14
A15467.03EA 59.6 EUR
A15467.03 A15467.06 A15467.14
4-(Trifluoromethoxy)benzenesulphonyl chloride ≥98%
4-(Trifluoromethoxy)benzenesulphonyl chloride

Purity: 98% CAS No.: 94108-56-2 Molecular Formula: C7H4ClF3O3S Formula Weight: 260.62 Flash Point: 110°(230°F) Refractive Index: 1.4800 Boiling Point: 120-122°/15mm Beilstein Reference: 7641524 UN No.: 3265 MDL No.: MFCD00042408

Kaava: C₇H₄ClF₃O₃S
Kiehumispiste: 120…122 °C (15 mmHg)
Tiheys: 1,536
Leimahduspiste: >110 °C (230 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00042408
CAS-numero: 94108-56-2
EINECS: 302-397-9
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR