3-Tolualdehydi ≥97% stabiloitu

Toimittaja: Thermo Fisher Scientific
Varoitus


Stabilization: Stabilised with hydroquinone (HQ)

A15313.14 A15313.06 A15313.22
A15313.14EA 138 EUR
A15313.14 A15313.06 A15313.22
3-Tolualdehydi ≥97% stabiloitu

Stabilized with 0.1% hydroquinone


Purity: 97% CAS No.: 620-23-5 Molecular Formula: C8H8O Formula Weight: 120.15 Flash Point: 78°(172°F) Refractive Index: 1.5410 Odour: Almond-like Boiling Point: 198-200° Beilstein Reference: 741964 MDL No.: MFCD00003374

Kaava: C₈H₈O
Kiehumispiste: 198…200 °C
Tiheys: 1,020
Leimahduspiste: 78 °C (172 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003374
CAS-numero: 620-23-5
EINECS: 210-632-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR