ω,3-Dibromiasetofenoni ≥97%

Toimittaja: Thermo Fisher Scientific
Vaara

A15210.06 A15210.14
A15210.06EA 182 EUR
A15210.06 A15210.14
ω,3-Dibromiasetofenoni ≥97%
ω,3-Dibromiasetofenoni

Purity: 97% CAS No.: 18523-22-3 Molecular Formula: C8H6Br2O Formula Weight: 277.95 Storage Temperature: 2°C to 8°C Odour: Pungent Beilstein Reference: 775786 UN No.: 3261 MDL No.: MFCD00082698

Kaava: C₈H₆Br₂O
Sulamispiste: 49…52 °C
MDL numero: MFCD00082698
CAS-numero: 18523-22-3
UN: 3261
ADR: 8,II

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR