3,4,5-Trimethoxytoluene ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

A15201.06 A15201.14 A15201.18
A15201.06EA 43.7 EUR
A15201.06 A15201.14 A15201.18
3,4,5-Trimethoxytoluene ≥98%
3,4,5-Trimethoxytoluene

Purity: 98% CAS No.: 6443-69-2 Molecular Formula: C10H14O3 Formula Weight: 182.22 Flash Point: 74°(165°F) Refractive Index: 1.5230 Melting Point: 34-38° Boiling Point: 115-116°/4mm Beilstein Reference: 1953791 MDL No.: MFCD00008397

Kaava: C₁₀H₁₄O₃
Kiehumispiste: 115…116 °C (4 mmHg)
Sulamispiste: 34…38 °C
Tiheys: 1,082
Leimahduspiste: 74 °C (165 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008397
CAS-numero: 6443-69-2
EINECS: 229-239-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR