2-jodipropaani ≥98% stabiloitu

Toimittaja: Thermo Fisher Scientific
Varoitus


Stabilization: Stabilised with copper

A15066.36 A15066.14 A15066.22
A15066.36EA 352 EUR
A15066.36 A15066.14 A15066.22
2-jodipropaani ≥98% stabiloitu
2-jodipropaani

Stabilized with copper


Purity: 98+% CAS No.: 75-30-9 Molecular Formula: (CH3)2CHI Formula Weight: 169.99 Flash Point: 42°(107°F) Physical Form: Liquid Density: 1.701 Refractive Index: 1.4982 Sensitivity: Light Sensitive Odour: Recognizable Melting Point: -90° Boiling Point: 88-90° Beilstein Reference: 1098244 Merck Reference: 14,5214 UN No.: 2392 MDL No.: MFCD00001071

Kaava: C₃H₇I
Kiehumispiste: 88…90 °C
Sulamispiste: –90 °C
Tiheys: 1,701
Leimahduspiste: 42 °C (107 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00001071
CAS-numero: 75-30-9
EINECS: 200-859-3
UN: 2392
ADR: 3,III
Merck Index: 14,5214

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR