(S)-(+)-Mandelic acid ≥99%

Toimittaja: Thermo Fisher Scientific
Vaara

A15061.06 A15061.14 A15061.22
A15061.06EA 63.9 EUR
A15061.06 A15061.14 A15061.22
(S)-(+)-Mandelic acid ≥99%
(S)-(+)-Mandelic acid

Purity: 99+% CAS No.: 17199-29-0 Molecular Formula: C8H8O3 Formula Weight: 152.15 Melting Point: 131-135° Beilstein Reference: 2208678

Kaava: C₈H₈O₃
Kiehumispiste: 322 °C (1013 hPa)
Sulamispiste: 131…135 °C
Tiheys: 1,349 g/cm³ (20 °C)
Leimahduspiste: >190 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004495
CAS-numero: 17199-29-0
EINECS: 241-240-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR