Isopropyyliamiini ≥99%

Toimittaja: Thermo Fisher Scientific
Vaara

A15044.0F A15044.AE A15044.AP
A15044.0FEA 233 EUR
A15044.0F A15044.AE A15044.AP
Isopropyyliamiini ≥99%
Isopropyyliamiini

Purity: 99+% CAS No.: 75-31-0 Molecular Formula: C3H9N Formula Weight: 59.11 Flash Point: -32°(-25°F) Physical Form: Clear liquid Density: 0.691 Refractive Index: 1.3750 Sensitivity: Air Sensitive Appearance: Colourless Odour: Amine-like Melting Point: -101° Boiling Point: 33-34° Beilstein Reference: 605259 Merck Reference: 14,5209 UN No.: 1221 MDL No.: MFCD00008082

Kaava: C₃H₉N
Kiehumispiste: 33…34 °C
Sulamispiste: –101 °C
Tiheys: 0,691
Leimahduspiste: –32 °C (–25 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008082
CAS-numero: 75-31-0
EINECS: 200-860-9
UN: 1221
ADR: 3,I
Merck Index: 14,5209

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR