Ethyl levulinate ≥98%

Toimittaja: Thermo Fisher Scientific

A15001.18 A15001.22 A15001.36
A15001.18EA 65.7 EUR
A15001.18 A15001.22 A15001.36
Ethyl levulinate ≥98%
Ethyl levulinate

Purity: 98% CAS No.: 539-88-8 Molecular Formula: C7H12O3 Formula Weight: 144.17 Flash Point: 94°(201°F) Refractive Index: 1.4220 Boiling Point: 203-205° Beilstein Reference: 507641 Merck Reference: 14,3819 MDL No.: MFCD00009209

Kaava: C₇H₁₂O₃
Kiehumispiste: 203…205 °C
Tiheys: 1,014
Leimahduspiste: 94 °C (201 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00009209
CAS-numero: 539-88-8
EINECS: 208-728-2
Merck Index: 14,3819

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR