(Chloromethyl)dimethylchlorosilane ≥98%

Toimittaja: Thermo Fisher Scientific
Vaara

A14899.09 A14899.14 A14899.22
A14899.09EA 87.3 EUR
A14899.09 A14899.14 A14899.22
(Chloromethyl)dimethylchlorosilane ≥98%
(Chloromethyl)dimethylchlorosilane

Purity: 98% CAS No.: 1719-57-9 Molecular Formula: C3H8Cl2Si Formula Weight: 143.09 Flash Point: 21°(69°F) Refractive Index: 1.4370 Odour: Acrid Boiling Point: 110-113° Beilstein Reference: 1731933 UN No.: 2985 MDL No.: MFCD00000875

Kaava: C₃H₈Cl₂Si
Kiehumispiste: 110…113 °C
Tiheys: 1,082
Leimahduspiste: 21 °C (69 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00000875
CAS-numero: 1719-57-9
EINECS: 217-006-6
UN: 2985
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR