Acetonyltriphenylphosphonium chloride ≥99%

Toimittaja: Thermo Fisher Scientific
Varoitus

A14691.14 A14691.22 A14691.36
A14691.14EA 73.3 EUR
A14691.14 A14691.22 A14691.36
Acetonyltriphenylphosphonium chloride ≥99%
Acetonyltriphenylphosphonium chloride

Purity: 99% CAS No.: 1235-21-8 Molecular Formula: C21H20ClOP Formula Weight: 354.82 Melting Point: 244-248° MDL No.: MFCD00011813

Kaava: C₂₁H₂₀ClOP
Sulamispiste: 244…248 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011813
CAS-numero: 1235-21-8
EINECS: 214-974-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR