Kadmiumsulfidi 98

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: Cadmium (II) sulphide

A14544.22 A14544.0E A14544.36
A14544.22EA 68.4 EUR
A14544.22 A14544.0E A14544.36
Kadmiumsulfidi 98
Kadmiumsulfidi

Purity: 98% CAS No.: 1306-23-6 Molecular Formula: CdS Formula Weight: 144.46 Odour: Odourless Melting Point: 980° subl. UN No.: 3077 MDL No.: MFCD00010922

Kaava: CdS
MW: 144.48 g/mol
Sulamispiste: 980 °C (subl.)
Tiheys: 4.82
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010922
CAS-numero: 1306-23-6
EINECS: 215-147-8
UN: 3077
ADR: 9 III
Merck Index: 12,01666

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR