Lawesson's reagent 97%

Toimittaja: Thermo Fisher Scientific
Vaara

A14530.06 A14530.14 A14530.22 A14530.36
A14530.06EA 48.9 EUR
A14530.06 A14530.14 A14530.22 A14530.36
Lawesson's reagent 97%
Lawesson's reagent

Purity: 97% CAS No.: 19172-47-5 Molecular Formula: C14H14O2P2S4 Formula Weight: 404.47 Melting Point: 221-225° Beilstein Reference: 1024888 UN No.: UN3134 MDL No.: MFCD00005171

Kaava: C₁₄H₁₄O₂P₂S₄
Sulamispiste: 221…225 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00005171
CAS-numero: 19172-47-5
EINECS: 242-855-4
ADR: 4.3,III
Merck Index: 14,5391

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR