2-Methyl-9,10-anthraquinone ≥97%

Toimittaja: Thermo Fisher Scientific

A14502.0B A14502.18 A14502.30
A14502.0BEA 826 EUR
A14502.0B A14502.18 A14502.30
2-Methyl-9,10-anthraquinone ≥97%
2-Methyl-9,10-anthraquinone

Purity: 97% CAS No.: 84-54-8 Molecular Formula: C15H10O2 Formula Weight: 222.24 Flash Point: 209°(408°F) Melting Point: 170-174° Beilstein Reference: 2050523 Merck Reference: 14,6021 MDL No.: MFCD00001235

Kaava: C₁₅H₁₀O₂
Kiehumispiste: 236…238 °C (10 mmHg)
Sulamispiste: 170…174 °C
Tiheys: 1,365 g/cm³ (20 °C)
Leimahduspiste: 209 °C (408 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00001235
CAS-numero: 84-54-8
EINECS: 201-539-6
UN: 3077
ADR: 9,III
Merck Index: 14,6021

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR